This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935554_m1
See other NEK4 GE Assays ›
Gene Symbol:
NEK4
Gene Name:
NIMA related kinase 4
Gene Aliases:
NRK2, STK2, pp12301
Chromosome Location:
Chr.3: 52707936 - 52770949 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935554_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEK4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193533.1 NP_001180462.1 11 - 12 1936 77
NM_003157.4 NP_003148.2 12 - 13 2203 77
XM_006713310.2 XP_006713373.1 11 - 12 2062 77
XM_011534039.2 XP_011532341.1 12 - 13 2200 77
XM_017007085.1 XP_016862574.1 12 - 13 2200 77
XM_017007086.1 XP_016862575.1 11 - 12 2062 77
GenBank mRNA AK294165.1 - 11 - 12 1843 77
AK312420.1 - 12 - 13 2191 77
BC063044.1 - 11 - 12 2055 5169184 77
EF560744.1 - 12 - 13 2199 77
KJ592714.1 - 11 - 12 1862 77
L20321.1 - 12 - 13 2178 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEK4

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
protein phosphorylation
cellular response to DNA damage stimulus
mitotic nuclear division
signal transduction by protein phosphorylation
positive regulation of transcription, DNA-templated
cell division
regulation of replicative cell aging
regulation of cellular senescence
regulation of response to DNA damage stimulus
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
manganese ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0