This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04421293_ft
See other CCDC6-RET FUSION GE Assays ›
Gene Symbol:
CCDC6-RET fusion CCDC6-RET fusion
Gene Name:
FUSION
FUSION
Gene Aliases:
CCDC6, RET
CCDC6, RET
Chromosome Location:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs04421293_ft
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCDC6-RET fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB698668.1 - 335 120
AB698669.1 - 335 120
AF395885.1 - 232 237
OM_CCDC6-RET_C1R12_COSF1271.0 - 105 120
Gene Symbol: CCDC6-RET fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA OM_CCDC6-RET_C1R12.0 - 79 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Fusion Homo sapiens

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000340058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000355710.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000479913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000498820.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 ENST00000615310.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020630.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007403.hg.1 NM_020975.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0