This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04409052_s1
See other C3ORF67-AS1 GE Assays ›
Gene Symbol:
C3orf67-AS1
Gene Name:
C3orf67 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 58824471 - 59019093 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04409052_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C3orf67-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110820.1 3 1220 103
GenBank mRNA AK090895.1 - 3 1220 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome 3 open reading frame 67 C3orf67 Anti-sense Chr. 3 - 58716415 - 59050078 on NCBI Build GRCh38
Downstream hCG1813818 LOC339902 618.2kb Anti-sense Chr. 3 - 59640163 - 59641872 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007625.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155192.hg.1 TC0300007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155196.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155196.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155196.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155196.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155196.hg.1 TC0300007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000493123.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000463703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000492031.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 ENST00000493123.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155189.hg.1 TC0300007625.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 ENST00000493123.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 ENST00000482372.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 ENST00000493123.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 NR_110820.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155195.hg.1 TC0300007625.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0