Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000604481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
ENST00000604481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241668.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241668.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241668.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241668.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241668.hg.1 |
TC0100018423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000604481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
ENST00000604481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018423.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241667.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241667.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241667.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241667.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241667.hg.1 |
TC0100018423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241666.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241666.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241666.hg.1 |
ENST00000566551.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241666.hg.1 |
NR_110616.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241666.hg.1 |
TC0100018423.hg.1 |
Clariom D TC |