Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008280.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158849.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158849.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158849.hg.1 |
TC0300008280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158862.hg.1 |
TC0300008280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158863.hg.1 |
TC0300008280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
ENST00000607746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
NR_110028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
NR_110030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158864.hg.1 |
TC0300008280.hg.1 |
Clariom D TC |