This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04408414_m1
See other DISC1FP1 GE Assays ›
Gene Symbol:
DISC1FP1
Gene Name:
DISC1 fusion partner 1 (non-protein coding)
Gene Aliases:
Boymaw
Chromosome Location:
Chr.11: 90251232 - 90915052 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04408414_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISC1FP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104190.1 6 - 7 588 72
GenBank mRNA CK000409.1 - 6 - 7 595 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 4490 MIR4490 -296.8kb Anti-sense Chr. 11 - 90555774 - 90555857 on NCBI Build GRCh38
Downstream FAT atypical cadherin 3 FAT3 CDHF15,
CDHR10,
hFat3
1.4mb Sense Chr. 11 - 92224636 - 92896473 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100008740.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164557.hg.1 TC1100008740.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164558.hg.1 TC1100008740.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000561596.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562245.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000562678.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000563681.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 ENST00000569513.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq
Affymetrix Human Clariom D Assay TC1100008740.hg.1 NR_104190.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0