Assay Details
Gene Symbol: DISC1FP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104190.1 | 5 - 6 | 503 | 110 | ||
GenBank mRNA | AB593165.1 | - | 5 - 6 | 428 | 110 | |
BM689797.1 | - | 2 - 3 | 144 | 110 | ||
CK000409.1 | - | 5 - 6 | 510 | 110 | ||
EU302123.1 | - | 5 - 6 | 450 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4490 | MIR4490 | -289.0kb | Anti-sense | Chr. 11 - 90555774 - 90555857 on NCBI Build GRCh38 | |
Downstream | microRNA 1261 | MIR1261 |
MIRN1261, hsa-mir-1261 |
16.4kb | Anti-sense | Chr. 11 - 90869121 - 90869202 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008740.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164550.hg.1 | TC1100008740.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000561596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000562678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000563681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | ENST00000569513.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | NR_104190.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164549.hg.1 | TC1100008740.hg.1 | Clariom D TC |
