Assay Details
Gene Symbol: FLG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103778.1 | 2 - 3 | 913 | 101 | ||
GenBank mRNA | AK056431.1 | - | 2 - 3 | 912 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hornerin | HRNR |
FLG3, S100A16, S100a18 |
-90.4kb | Anti-sense | Chr. 1 - 152212076 - 152224196 on NCBI Build GRCh38 |
Downstream | filaggrin family member 2 | FLG2 | IFPS | 16.0kb | Anti-sense | Chr. 1 - 152348735 - 152360028 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018291.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239427.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239428.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239435.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239422.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239423.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239431.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239421.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | ENST00000629331.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239434.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | ENST00000445097.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | NR_103779.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239432.hg.1 | TC0100018291.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | ENST00000392688.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | NR_103778.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239425.hg.1 | TC0100018291.hg.1 | Clariom D TC |
