This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407940_m1
See other GHRLOS GE Assays ›
Gene Symbol:
GHRLOS
Gene Name:
ghrelin opposite strand/antisense RNA
Gene Aliases:
GHRL-AS1, GHRLAS, NCRNA00068
Chromosome Location:
Chr.3: 10280952 - 10293449 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407940_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GHRLOS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004431.3 2 - 3 149 98
NR_024144.2 3 - 4 232 98
NR_073566.1 4 - 5 328 98
NR_073568.1 3 - 4 232 98
GenBank mRNA EF549568.1 - 1 - 2 92 98
EF549570.1 - 1 - 2 92 98
EU789535.1 - 3 - 4 231 98
EU789536.1 - 3 - 4 231 98
EU789538.1 - 2 - 3 148 98
EU789539.1 - 2 - 3 148 98
EU789541.1 - 2 - 3 148 98
EU789542.1 - 2 - 3 148 98
EU789543.1 - 4 - 5 327 98
EU789545.1 - 3 - 4 231 98
EU789546.1 - 2 - 3 148 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CF264800.1 - 1 - 2 58 Chr. 3 - 10285788 - 10292630 [+] on Build GRCh38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 852 LINC00852 C3orf42,
GHRL-AS2,
GHRLOS2,
NAG73
-2.0 Sense Chr. 3 - 10284419 - 10285746 on NCBI Build GRCh38
Downstream SEC13 homolog,
nuclear pore and COPII coat complex component
SEC13 D3S1231E,
SEC13L1,
SEC13R,
npp-20
13.4kb Anti-sense Chr. 3 - 10300929 - 10321188 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0