Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
ENST00000602736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
ENST00000602736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016639.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225828.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225828.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225828.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225828.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225828.hg.1 |
TC0200016639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200225827.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225827.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225827.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225827.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225827.hg.1 |
TC0200016639.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200225829.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225829.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225829.hg.1 |
ENST00000449391.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200225829.hg.1 |
NR_046836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200225829.hg.1 |
TC0200016639.hg.1 |
Clariom D TC |