This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407709_m1
See other HTR2A-AS1 GE Assays ›
Gene Symbol:
HTR2A-AS1
Gene Name:
HTR2A antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 46852140 - 46856299 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407709_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HTR2A-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046612.1 2 - 3 227 113
GenBank mRNA AI914390.1 - 1 - 2 113

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream 5-hydroxytryptamine receptor 2A HTR2A 5-HT2A,
HTR2
Anti-sense Chr. 13 - 46831542 - 46897076 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 562 LINC00562 1.1mb Anti-sense Chr. 13 - 47930153 - 47932622 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300007115.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 NR_103752.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007115.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148232.hg.1 TC1300007115.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148236.hg.1 TC1300007115.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 ENST00000455126.1 Ensembl
Affymetrix Human Clariom D Assay TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay TC1300007115.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay TC1300007115.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay TC1300007115.hg.1 NR_103752.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000430913.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 ENST00000452352.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 NR_046612.1 Refseq
Affymetrix Human Clariom D Assay PSR1300148233.hg.1 TC1300007115.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0