This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407259_m1
See other BHLHE40-AS1 GE Assays ›
Gene Symbol:
BHLHE40-AS1
Gene Name:
BHLHE40 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 4896809 - 4979961 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407259_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BHLHE40-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125915.1 3 - 4 466 97
NR_125916.1 2 - 3 326 97
GenBank mRNA AK056892.1 - 3 - 4 466 97
DA948054.1 - 2 - 3 273 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA948464.1 - 2 - 3 274 Chr. 3 - 4898417 - 4979908 [-] on Build GRCh38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream inositol 1,4,5-trisphosphate receptor type 1 ITPR1 ACV,
CLA4,
INSP3R1,
IP3R,
IP3R1,
PPP1R94,
SCA15,
SCA16,
SCA29
-49.0kb Anti-sense Chr. 3 - 4493348 - 4847840 on NCBI Build GRCh38
Downstream basic helix-loop-helix family member e40 BHLHE40 BHLHB2,
DEC1,
HLHB2,
SHARP-2,
STRA13,
Stra14
78.7kb Anti-sense Chr. 3 - 4979412 - 4985181 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300010139.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000441386.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay TC0300010139.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173889.hg.1 TC0300010139.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173890.hg.1 TC0300010139.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173892.hg.1 TC0300010139.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000420832.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000434530.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000438479.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000615017.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000615178.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 ENST00000620618.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_037903.3 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125914.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 NR_125916.1 Refseq
Affymetrix Human Clariom D Assay PSR0300173893.hg.1 TC0300010139.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0