This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407126_m1
See other LOC102723729 GE Assays ›
Gene Symbol:
LOC102723729
Gene Name:
uncharacterized LOC102723729
Gene Aliases:
-
Chromosome Location:
Chr.8: 41534305 - 41578368 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407126_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102723729
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125824.1 2 - 3 305 82
GenBank mRNA AI702202.1 - 2 - 3 305 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW204780.1 - 2 - 3 304 Chr. 8 - 41534320 - 41578367 [-] on Build GRCh38 82
GenBankmRNA BE218154.1 - 2 - 3 305 Chr. 8 - 41534155 - 41578368 [-] on Build GRCh38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream GINS complex subunit 4 GINS4 SLD5 Anti-sense Chr. 8 - 41529206 - 41545046 on NCBI Build GRCh38
Downstream glycerol-3-phosphate acyltransferase 4 GPAT4 1-AGPAT 6,
AGPAT6,
LPAAT-zeta,
LPAATZ,
TSARG7
40.9kb Anti-sense Chr. 8 - 41578188 - 41625001 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010244.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010244.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay TC0800010244.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay PSR0800169673.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800169673.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay PSR0800169673.hg.1 ENST00000523081.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800169673.hg.1 NR_125824.1 Refseq
Affymetrix Human Clariom D Assay PSR0800169673.hg.1 TC0800010244.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0