This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407006_m1
See other CEBPB-AS1 GE Assays ›
Gene Symbol:
CEBPB-AS1
Gene Name:
CEBPB antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 50184603 - 50192069 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407006_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPB-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125739.1 2 - 3 566 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 1273 LINC01273 -13.3kb Anti-sense Chr. 20 - 50172570 - 50176671 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1272 LINC01272 76.1kb Anti-sense Chr. 20 - 50267468 - 50279795 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009418.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165904.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000165904.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165904.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000165904.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165904.hg.1 TC2000009418.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000165907.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165907.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165907.hg.1 TC2000009418.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000165905.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000165905.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165905.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000165905.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay PSR2000165905.hg.1 TC2000009418.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 ENST00000613921.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq
Affymetrix Human Clariom D Assay TC2000009418.hg.1 NR_125739.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0