This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406859_m1
See other BAHD1 GE Assays ›
Gene Symbol:
BAHD1
Gene Name:
bromo adjacent homology domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 40439721 - 40468242 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406859_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAHD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125372.1 3 - 4 357 96
XM_011521368.2 XP_011519670.2 5 - 6 2309 96
GenBank mRNA AL833923.1 - 4 - 5 1278 96
HY353002.1 - 2 - 3 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI196318.1 - 2 - 3 139 Chr. 15 - 40463968 - 40466247 [+] on Build GRCh38 96
GenBankmRNA BI767114.1 - 3 - 4 278 Chr. 15 - 40462256 - 40466090 [+] on Build GRCh38 96
GenBankmRNA CD626430.1 - 2 - 3 Chr. 15 - 40462272 - 40466039 [-] on Build GRCh38 96
GenBankmRNA CD626429.1 - 3 - 4 269 Chr. 15 - 40462272 - 40466039 [+] on Build GRCh38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream isovaleryl-CoA dehydrogenase IVD ACAD2 -28.5kb Sense Chr. 15 - 40405485 - 40435948 on NCBI Build GRCh38
Downstream carbohydrate sulfotransferase 14 CHST14 ATCS,
D4ST1,
EDSMC1,
HNK1ST
5.5kb Sense Chr. 15 - 40470961 - 40473158 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500006956.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing
transcription, DNA-templated
covalent chromatin modification
heterochromatin assembly
negative regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
chromatin binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000416165.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000560846.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000561234.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000561464.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NM_001301132.1 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NM_014952.4 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NR_125372.1 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000416165.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000560846.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000561234.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 ENST00000561464.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NM_001301132.1 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NM_014952.4 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 NR_125372.1 Refseq
Affymetrix Human Clariom D Assay PSR1500148451.hg.1 TC1500006956.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0