This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406711_m1
See other BMPR1B-AS1 GE Assays ›
Gene Symbol:
BMPR1B-AS1
Gene Name:
BMPR1B antisense RNA 1 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.4: 94743668 - 94757533 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406711_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BMPR1B-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_121610.1 1 - 2 141 98
GenBank mRNA AI150588.1 - 2 - 3 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PDZ and LIM domain 5 PDLIM5 ENH,
ENH1,
L9,
LIM
-86.6kb Anti-sense Chr. 4 - 94451857 - 94668227 on NCBI Build GRCh38
Downstream bone morphogenetic protein receptor type 1B BMPR1B ALK-6,
ALK6,
AMDD,
BDA1D,
BDA2,
CDw293
444.0 Anti-sense Chr. 4 - 94757977 - 95158453 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011353.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177617.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400177617.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177617.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400177617.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177617.hg.1 TC0400011353.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011353.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay TC0400011353.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177616.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400177616.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177616.hg.1 ENST00000510795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400177616.hg.1 NR_121610.1 Refseq
Affymetrix Human Clariom D Assay PSR0400177616.hg.1 TC0400011353.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0