This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406439_g1
See other LINC01163 GE Assays ›
Gene Symbol:
LINC01163
Gene Name:
long intergenic non-protein coding RNA 1163
Gene Aliases:
TCONS_00017722
Chromosome Location:
Chr.10: 128285950 - 128317726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406439_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120619.1 4 - 5 1917 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream marker of proliferation Ki-67 MKI67 KIA,
MIB-,
MIB-1,
PPP1R105
-188.1kb Anti-sense Chr. 10 - 128096659 - 128126405 on NCBI Build GRCh38
Downstream O-6-methylguanine-DNA methyltransferase MGMT 1.2mb Sense Chr. 10 - 129467184 - 129770983 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009303.hg.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000626518.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay TC1000009303.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163767.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000163767.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163767.hg.1 ENST00000630252.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000163767.hg.1 NR_120619.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163767.hg.1 TC1000009303.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0