Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163764.hg.1 |
TC1000009303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000163765.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163765.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163765.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163765.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163765.hg.1 |
TC1000009303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000626518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
ENST00000630252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009303.hg.1 |
NR_120619.1 |
Refseq |