This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406264_m1
See other LOC100506675 GE Assays ›
Gene Symbol:
LOC100506675
Gene Name:
uncharacterized LOC100506675
Gene Aliases:
-
Chromosome Location:
Chr.11: 32035979 - 32041318 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406264_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100506675
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120527.1 1 - 2 209 108
GenBank mRNA BX099517.1 - 1 - 2 173 108
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW182974.1 - 1 - 2 Chr. 11 - 32040065 - 32041248 [+] on Build GRCh38 108
GenBankmRNA DB024222.1 - 1 - 2 182 Chr. 11 - 32040000 - 32041291 [-] on Build GRCh38 108
GenBankmRNA AW183760.1 - 1 - 2 Chr. 11 - 32040112 - 32041172 [+] on Build GRCh38 108
GenBankmRNA AI027739.1 - 1 - 2 Chr. 11 - 32040107 - 32041214 [+] on Build GRCh38 108
GenBankmRNA AI936632.1 - 1 - 2 Chr. 11 - 32040112 - 32041253 [+] on Build GRCh38 108
GenBankmRNA AI808144.1 - 1 - 2 Chr. 11 - 32040065 - 32041231 [+] on Build GRCh38 108

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC440034 DKFZp686K1684 -153.0kb Anti-sense Chr. 11 - 31816566 - 31887041 on NCBI Build GRCh38
Downstream reticulocalbin 1 RCN1 HEL-S-84,
PIG20,
RCAL,
RCN
49.6kb Anti-sense Chr. 11 - 32090931 - 32105726 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0