Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159194.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159194.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159194.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159194.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159194.hg.1 |
TC0600008510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159192.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159192.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159192.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159192.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159192.hg.1 |
TC0600008510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159193.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159193.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159193.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159193.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159193.hg.1 |
TC0600008510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159195.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159195.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159195.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159195.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159195.hg.1 |
TC0600008510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
ENST00000445310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008510.hg.1 |
NR_120503.1 |
Refseq |