Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007947.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156956.hg.1 |
TC1500007947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
ENST00000557799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
NR_120361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
NR_120362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156957.hg.1 |
TC1500007947.hg.1 |
Clariom D TC |