This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405847_m1
See other LINGO1-AS2 GE Assays ›
Gene Symbol:
LINGO1-AS2
Gene Name:
LINGO1 antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 77660051 - 77668074 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405847_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINGO1-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120361.1 1 - 2 134 95
GenBank mRNA BC038758.1 - 1 - 2 133 95
BI463387.1 - 1 - 2 138 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leucine rich repeat and Ig domain containing 1 LINGO1 LERN1,
LRRN6A,
UNQ201
Anti-sense Chr. 15 - 77613024 - 77820900 on NCBI Build GRCh38
Downstream golgin A6 family member A pseudogene LOC645752 247.1kb Anti-sense Chr. 15 - 77914217 - 77926846 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500007947.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay PSR1500156959.hg.1 TC1500007947.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007947.hg.1 ENST00000557799.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120361.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120362.1 Refseq
Affymetrix Human Clariom D Assay TC1500007947.hg.1 NR_120362.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0