This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405760_m1
See other LINC01359 GE Assays ›
Gene Symbol:
LINC01359
Gene Name:
long intergenic non-protein coding RNA 1359
Gene Aliases:
-
Chromosome Location:
Chr.1: 64979577 - 65002476 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405760_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01359
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_119383.1 4 - 5 455 84
GenBank mRNA BC035370.1 - 4 - 5 455 84
BI825893.1 - 4 - 5 455 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI826506.1 - 4 - 5 456 Chr. 1 - 64987461 - 65002475 [-] on Build GRCh38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Janus kinase 1 JAK1 JAK1A,
JAK1B,
JTK3
-20.4kb Sense Chr. 1 - 64833223 - 65067746 on NCBI Build GRCh38
Downstream microRNA 3671 MIR3671 68.3kb Sense Chr. 1 - 65057755 - 65057842 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014459.hg.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay PSR0100208177.hg.1 TC0100014459.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay PSR0100208175.hg.1 TC0100014459.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000415842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000436350.2 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000444349.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 ENST00000447748.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq
Affymetrix Human Clariom D Assay TC0100014459.hg.1 NR_119383.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0