Assay Details
Gene Symbol: ADGRA1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111905.1 | 1 - 2 | 216 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adhesion G protein-coupled receptor A1 | ADGRA1 | GPR123 | Anti-sense | Chr. 10 - 133070929 - 133131675 on NCBI Build GRCh38 | |
Downstream | kinase non-catalytic C-lobe domain containing 1 | KNDC1 |
C10orf23, RASGEF2, bB439H18.3 |
73.8kb | Anti-sense | Chr. 10 - 133160096 - 133226412 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184447.hg.1 | TC1000012378.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012378.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184444.hg.1 | TC1000012378.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000366099.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | ENST00000444433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | NR_111905.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184448.hg.1 | TC1000012378.hg.1 | Clariom D TC |
