This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405342_m1
See other ITFG1-AS1 GE Assays ›
Gene Symbol:
ITFG1-AS1
Gene Name:
ITFG1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 47144068 - 47162747 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405342_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ITFG1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110903.1 4 - 5 689 100
NR_110904.1 3 - 4 524 100
GenBank mRNA BE466653.1 - 1 - 2 100
BF115500.1 - 1 - 2 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA730133.1 - 1 - 2 Chr. 16 - 47144380 - 47162697 [-] on Build GRCh38 100
GenBankmRNA AI377784.1 - 1 - 2 Chr. 16 - 47144360 - 47162698 [-] on Build GRCh38 100
GenBankmRNA AI221448.1 - 1 - 2 Chr. 16 - 47144473 - 47162710 [-] on Build GRCh38 100
GenBankmRNA HY227067.1 - 1 - 2 Chr. 16 - 47144446 - 47162691 [-] on Build GRCh38 100
GenBankmRNA HY239209.1 - 1 - 2 Chr. 16 - 47147387 - 47162691 [-] on Build GRCh38 100
GenBankmRNA AI611624.1 - 1 - 2 Chr. 16 - 47144408 - 47162697 [-] on Build GRCh38 100
GenBankmRNA AI936568.1 - 1 - 2 Chr. 16 - 47144313 - 47162747 [-] on Build GRCh38 100
GenBankmRNA HY239328.1 - 1 - 2 Chr. 16 - 47144282 - 47162691 [-] on Build GRCh38 100
GenBankmRNA BE551587.1 - 1 - 2 Chr. 16 - 47144282 - 47162697 [-] on Build GRCh38 100
GenBankmRNA AW303619.1 - 1 - 2 Chr. 16 - 47144272 - 47162747 [-] on Build GRCh38 100
GenBankmRNA AI147974.1 - 1 - 2 Chr. 16 - 47157550 - 47162698 [-] on Build GRCh38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neuropilin and tolloid like 2 NETO2 BTCL2,
NEOT2
-17.5kb Anti-sense Chr. 16 - 47081520 - 47144025 on NCBI Build GRCh38
Downstream phosphorylase kinase regulatory subunit beta PHKB 298.5kb Sense Chr. 16 - 47461131 - 47701523 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600007733.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156403.hg.1 TC1600007733.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156404.hg.1 TC1600007733.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0