This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405340_m1
See other ITFG1-AS1 GE Assays ›
Gene Symbol:
ITFG1-AS1
Gene Name:
ITFG1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 47144068 - 47162747 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405340_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ITFG1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110903.1 2 - 3 598 139
GenBank mRNA BF115500.1 - 3 - 4 139
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY227067.1 - 3 - 4 Chr. 16 - 47144446 - 47162691 [-] on Build GRCh38 139
GenBankmRNA HY239209.1 - 3 - 4 Chr. 16 - 47147387 - 47162691 [-] on Build GRCh38 139

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neuropilin and tolloid like 2 NETO2 BTCL2,
NEOT2
-9.6kb Anti-sense Chr. 16 - 47081520 - 47144025 on NCBI Build GRCh38
Downstream phosphorylase kinase regulatory subunit beta PHKB 303.7kb Sense Chr. 16 - 47461131 - 47701523 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600007733.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay TC1600007733.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000564705.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 ENST00000565694.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 NR_110903.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 NR_110904.1 Refseq
Affymetrix Human Clariom D Assay PSR1600156401.hg.1 TC1600007733.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0