This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405240_m1
See other EDRF1 GE Assays ›
Gene Symbol:
EDRF1
Gene Name:
erythroid differentiation regulatory factor 1
Gene Aliases:
C10orf137
Chromosome Location:
Chr.10: 125719515 - 125764143 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405240_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EDRF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110858.1 18 - 19 2795 97
GenBank mRNA BC026172.1 - 18 - 19 2796 4838438 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DR760365.1 - 1 - 2 137 Chr. 10 - 125742223 - 125748001 [+] on Build GRCh38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC399821 FLJ37035 -22.7kb Anti-sense Chr. 10 - 125705290 - 125719493 on NCBI Build GRCh38
Downstream EDRF1 antisense RNA 1 EDRF1-AS1 1.5kb Anti-sense Chr. 10 - 125744727 - 125752060 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009240.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000337623.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000356792.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000368815.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000419769.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000481600.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000525091.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NM_001202438.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NM_015608.2 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110857.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110858.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110859.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000337623.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000356792.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000368815.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000419769.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000481600.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 ENST00000525091.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NM_001202438.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NM_015608.2 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110857.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110858.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 NR_110859.1 Refseq
Affymetrix Human Clariom D Assay PSR1000163372.hg.1 TC1000009240.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0