This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405160_m1
See other PRKCA-AS1 GE Assays ›
Gene Symbol:
PRKCA-AS1
Gene Name:
PRKCA antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 66398069 - 66416854 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405160_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKCA-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110822.1 3 - 4 1201 68
GenBank mRNA AI201109.1 - 1 - 2 68
BC033554.1 - 2 - 3 1959 68
BG183684.1 - 1 - 2 78 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG216420.1 - 1 - 2 79 Chr. 17 - 66398069 - 66412764 [-] on Build GRCh38 68
GenBankmRNA BG212263.1 - 1 - 2 79 Chr. 17 - 66398070 - 66412764 [-] on Build GRCh38 68
GenBankmRNA BG181623.1 - 1 - 2 108 Chr. 17 - 66398066 - 66412755 [-] on Build GRCh38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein kinase C alpha PRKCA AAG6,
PKC-alpha,
PKCA,
PRKACA
Anti-sense Chr. 17 - 66302640 - 66810744 on NCBI Build GRCh38
Downstream microRNA 634 MIR634 MIRN634,
hsa-mir-634
374.3kb Anti-sense Chr. 17 - 66787072 - 66787168 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011498.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000583421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000583421.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000583421.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000583421.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584614.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584614.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay TC1700011498.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 ENST00000584715.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 NR_110822.1 Refseq
Affymetrix Human Clariom D Assay PSR1700193147.hg.1 TC1700011498.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0