This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405136_m1
See other LOC100507002 GE Assays ›
Gene Symbol:
LOC100507002
Gene Name:
uncharacterized LOC100507002
Gene Aliases:
-
Chromosome Location:
Chr.17: 65100812 - 65111095 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405136_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100507002
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110801.1 2 - 3 313 101
GenBank mRNA DA745846.1 - 2 - 3 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ285910.1 - 1 - 2 Chr. 17 - 65100943 - 65111089 [-] on Build GRCh38 101
GenBankmRNA BQ269268.1 - 2 - 3 Chr. 17 - 65100849 - 65111027 [+] on Build GRCh38 101
GenBankmRNA BQ777364.1 - 2 - 3 Chr. 17 - 65100828 - 65111058 [+] on Build GRCh38 101
GenBankmRNA BE778082.1 - 2 - 3 Chr. 17 - 65100854 - 65111197 [+] on Build GRCh38 101
GenBankmRNA DB470502.1 - 2 - 3 Chr. 17 - 65100812 - 65110829 [+] on Build GRCh38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein subunit alpha 13 GNA13 G13 -44.4kb Anti-sense Chr. 17 - 65009289 - 65056802 on NCBI Build GRCh38
Downstream regulator of G-protein signaling 9 RGS9 PERRS,
RGS9L
26.2kb Sense Chr. 17 - 65137338 - 65227703 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008634.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164592.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164592.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164592.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164592.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164592.hg.1 TC1700008634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700164589.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164589.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164589.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164589.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164589.hg.1 TC1700008634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700164591.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164591.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164591.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164591.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164591.hg.1 TC1700008634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700164594.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164594.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164594.hg.1 TC1700008634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay TC1700008634.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164590.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164590.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164590.hg.1 ENST00000582940.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700164590.hg.1 NR_110801.1 Refseq
Affymetrix Human Clariom D Assay PSR1700164590.hg.1 TC1700008634.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0