This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404909_m1
See other LOC101927069 GE Assays ›
Gene Symbol:
LOC101927069
Gene Name:
uncharacterized LOC101927069
Gene Aliases:
-
Chromosome Location:
Chr.9: 68953088 - 68975841 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404909_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927069
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110647.1 2 - 3 357 72
GenBank mRNA AK057188.1 - 2 - 3 357 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA952460.1 - 2 - 3 359 Chr. 9 - 68954748 - 68975843 [-] on Build GRCh38 72
GenBankmRNA DA564081.1 - 2 - 3 359 Chr. 9 - 68954748 - 68975843 [-] on Build GRCh38 72
GenBankmRNA DA593275.1 - 2 - 3 330 Chr. 9 - 68954728 - 68975814 [-] on Build GRCh38 72
GenBankmRNA DA970473.1 - 2 - 3 357 Chr. 9 - 68954587 - 68975841 [-] on Build GRCh38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream family with sequence similarity 122A FAM122A C9orf42 -169.4kb Anti-sense Chr. 9 - 68780006 - 68783879 on NCBI Build GRCh38
Downstream protein kinase cAMP-activated catalytic subunit gamma PRKACG BDPLT19,
KAPG,
PKACg
55.3kb Sense Chr. 9 - 69012529 - 69014123 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010332.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0900170743.hg.1 NR_110647.1 Refseq
Affymetrix Human Clariom D Assay PSR0900170743.hg.1 NR_110647.1 Refseq
Affymetrix Human Clariom D Assay PSR0900170743.hg.1 TC0900010332.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900170742.hg.1 NR_110647.1 Refseq
Affymetrix Human Clariom D Assay PSR0900170742.hg.1 NR_110647.1 Refseq
Affymetrix Human Clariom D Assay PSR0900170742.hg.1 TC0900010332.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0