This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404715_m1
See other LANCL1-AS1 GE Assays ›
Gene Symbol:
LANCL1-AS1
Gene Name:
LANCL1 antisense RNA 1
Gene Aliases:
LANCLOT
Chromosome Location:
Chr.2: 210324712 - 210470333 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404715_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LANCL1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110604.1 1 - 2 121 80
NR_110605.1 1 - 2 121 80
NR_110606.1 1 - 2 121 80
GenBank mRNA AL599033.1 - 1 - 2 91 80
BX119066.1 - 1 - 2 67 80
DA820426.1 - 1 - 2 121 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA F25413.1 - 1 - 2 95 Chr. 2 - 210324744 - 210460516 [+] on Build GRCh38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream myosin light chain 1 MYL1 MLC1F,
MLC3F
-9.5kb Anti-sense Chr. 2 - 210290144 - 210315171 on NCBI Build GRCh38
Downstream LanC like 1 LANCL1 GPR69A,
p40
1.8kb Anti-sense Chr. 2 - 210431249 - 210477618 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010655.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000416344.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay TC0200010655.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175604.hg.1 TC0200010655.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000420418.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 ENST00000433296.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175601.hg.1 TC0200010655.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110604.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110605.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 NR_110606.1 Refseq
Affymetrix Human Clariom D Assay PSR0200175599.hg.1 TC0200010655.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0