This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404679_m1
See other ADGRL3-AS1 GE Assays ›
Gene Symbol:
ADGRL3-AS1
Gene Name:
adhesion G protein-coupled receptor L3 antisense RNA 1
Gene Aliases:
LPHN3-AS1
Chromosome Location:
Chr.4: 62071755 - 62161765 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404679_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110595.1 2 - 3 82 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB453279.1 - 3 - 4 221 Chr. 4 - 62072828 - 62165554 [-] on Build GRCh38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adhesion G protein-coupled receptor L3 ADGRL3 CIRL3,
CL3,
LEC3,
LPHN3
-58.3kb Anti-sense Chr. 4 - 61201256 - 62078335 on NCBI Build GRCh38
Downstream trans-2,3-enoyl-CoA reductase-like TECRL GPSN2L,
SRD5A2L2,
TERL
2.1mb Sense Chr. 4 - 64276298 - 64409509 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010832.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173640.hg.1 TC0400010832.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000502339.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 ENST00000509461.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay TC0400010832.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000504135.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 NR_110595.1 Refseq
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 ENST00000506704.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400173642.hg.1 TC0400010832.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0