This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404082_m1
See other UTAT33 GE Assays ›
Gene Symbol:
UTAT33
Gene Name:
uncharacterized LOC101927492
Gene Aliases:
-
Chromosome Location:
Chr.2: 105097052 - 105102944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404082_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UTAT33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110227.1 3 - 4 231 98
GenBank mRNA AI381531.1 - 1 - 2 98
CR736606.1 - 3 - 4 231 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY204814.1 - 1 - 2 Chr. 2 - 105097056 - 105098664 [+] on Build GRCh38 98
GenBankmRNA AA147483.1 - 3 - 4 231 Chr. 2 - 105097162 - 105102944 [-] on Build GRCh38 98
GenBankmRNA CR739107.1 - 3 - 4 231 Chr. 2 - 105097060 - 105102944 [-] on Build GRCh38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC102724691 LOC102724691 -58.6kb Sense Chr. 2 - 104936241 - 105038496 on NCBI Build GRCh38
Downstream uncharacterized LOC105373984 LOC105373984 44.7kb Anti-sense Chr. 2 - 105144113 - 105145424 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013741.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay PSR0200200714.hg.1 TC0200013741.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000432211.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 ENST00000449177.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay TC0200013741.hg.1 NR_110227.1 Refseq
Affymetrix Human Clariom D Assay TC0200013741.hg.1 NR_110227.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0