Assay Details
Gene Symbol: PHACTR2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110148.1 | 1 - 2 | 173 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
fucosidase, alpha-L- 2, plasma |
FUCA2 | dJ20N2.5 | -49.3kb | Sense | Chr. 6 - 143494812 - 143511883 on NCBI Build GRCh38 |
Downstream | phosphatase and actin regulator 2 | PHACTR2 | C6orf56 | 46.1kb | Anti-sense | Chr. 6 - 143608180 - 143831185 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192751.hg.1 | TC0600013428.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192748.hg.1 | TC0600013428.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | ENST00000450575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027113.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_027114.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110148.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013428.hg.1 | NR_110149.1 | Refseq |
