This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403429_m1
See other TARID GE Assays ›
Gene Symbol:
TARID
Gene Name:
TCF21 antisense RNA inducing promoter demethylation
Gene Aliases:
EYA4-AS1
Chromosome Location:
Chr.6: 133502252 - 133889006 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403429_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TARID
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109982.1 5 - 6 705 93
GenBank mRNA BC041459.1 - 5 - 6 681 93
DB279014.1 - 3 - 4 407 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream EYA transcriptional coactivator and phosphatase 4 EYA4 CMD1J,
DFNA10
-3.7kb Anti-sense Chr. 6 - 133240340 - 133532120 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1312 LINC01312 252.9kb Anti-sense Chr. 6 - 133821147 - 133853992 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013220.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0