This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403024_m1
See other DIAPH3-AS1 GE Assays ›
Gene Symbol:
DIAPH3-AS1
Gene Name:
DIAPH3 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 60012718 - 60044357 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403024_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIAPH3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109838.1 2 - 3 231 114
NR_109839.1 2 - 3 159 114
GenBank mRNA BC032915.2 - 2 - 3 162 114
BX398653.2 - 2 - 3 222 114
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE502507.1 - 1 - 2 Chr. 13 - 60020918 - 60025150 [-] on Build GRCh38 114
GenBankmRNA AI809360.1 - 1 - 2 Chr. 13 - 60020918 - 60025154 [-] on Build GRCh38 114
GenBankmRNA BX398652.2 - 3 - 4 Chr. 13 - 60012727 - 60044321 [-] on Build GRCh38 114
GenBankmRNA BI458826.1 - 2 - 3 167 Chr. 13 - 60013301 - 60025154 [+] on Build GRCh38 114
GenBankmRNA BX092220.1 - 2 - 3 188 Chr. 13 - 60013274 - 60025132 [+] on Build GRCh38 114

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101926897 LOC101926897 -1.8mb Sense Chr. 13 - 58165827 - 58209483 on NCBI Build GRCh38
Downstream DIAPH3 antisense RNA 2 DIAPH3-AS2 DIPAH3-AS2 119.8kb Sense Chr. 13 - 60144698 - 60153505 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300007335.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1300149681.hg.1 NR_109839.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149681.hg.1 NR_109839.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149681.hg.1 TC1300007335.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300149680.hg.1 NR_109838.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149680.hg.1 NR_109839.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149680.hg.1 NR_109838.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149680.hg.1 NR_109839.1 Refseq
Affymetrix Human Clariom D Assay PSR1300149680.hg.1 TC1300007335.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0