This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402853_g1
See other FAM83C-AS1 GE Assays ›
Gene Symbol:
FAM83C-AS1
Gene Name:
FAM83C antisense RNA 1
Gene Aliases:
C20orf120, NCRNA00154, dJ614O4.3
Chromosome Location:
Chr.20: 35285251 - 35285772 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402853_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM83C-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109815.1 1 - 2 66 83
GenBank mRNA AI214830.1 - 1 - 2 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream family with sequence similarity 83 member C FAM83C C20orf128,
dJ614O4.7
Anti-sense Chr. 20 - 35285731 - 35292422 on NCBI Build GRCh38
Downstream ubiquinol-cytochrome c reductase complex assembly factor 1 UQCC1 BFZB,
C20orf44,
CBP3,
UQCC
16.8kb Anti-sense Chr. 20 - 35302566 - 35412142 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007214.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007214.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007214.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007214.hg.1 NR_109815.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007214.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149581.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149581.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149581.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149581.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149581.hg.1 TC2000007214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000149582.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149582.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149582.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149582.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149582.hg.1 TC2000007214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2000007214.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007214.hg.1 ENST00000429167.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007214.hg.1 NR_109815.1 Refseq
Affymetrix Human Clariom D Assay TC2000007214.hg.1 NR_109815.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0