This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402431_m1
See other TPD52 GE Assays ›
Gene Symbol:
TPD52
Gene Name:
tumor protein D52
Gene Aliases:
D52, N8L, PC-1, PrLZ, hD52
Chromosome Location:
Chr.8: 80034868 - 80171659 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402431_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPD52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105036.1 1 - 2 178 89
GenBank mRNA AK308983.1 - 1 - 2 90 89
DA665049.1 - 1 - 2 178 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA518361.1 - 1 - 2 104 Chr. 8 - 80051525 - 80171126 [-] on Build GRCh38 89
GenBankmRNA HY152824.1 - 1 - 2 95 Chr. 8 - 80051527 - 80171116 [-] on Build GRCh38 89
GenBankmRNA DA284052.1 - 1 - 2 90 Chr. 8 - 80042618 - 80171110 [-] on Build GRCh38 89
GenBankmRNA DA181509.1 - 1 - 2 106 Chr. 8 - 80044183 - 80171128 [-] on Build GRCh38 89
GenBankmRNA DA254900.1 - 1 - 2 179 Chr. 8 - 80051527 - 80171200 [-] on Build GRCh38 89
GenBankmRNA HY109086.1 - 1 - 2 95 Chr. 8 - 80038216 - 80171116 [-] on Build GRCh38 89
GenBankmRNA DB013663.1 - 1 - 2 178 Chr. 8 - 80051530 - 80171200 [-] on Build GRCh38 89
GenBankmRNA HY137904.1 - 1 - 2 95 Chr. 8 - 80051538 - 80171116 [-] on Build GRCh38 89
GenBankmRNA HY119008.1 - 1 - 2 95 Chr. 8 - 80042653 - 80171116 [-] on Build GRCh38 89
GenBankmRNA DA246080.1 - 1 - 2 99 Chr. 8 - 80042616 - 80171121 [-] on Build GRCh38 89
GenBankmRNA BU429623.1 - 144 Chr. - - [-] on Build 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mitochondrial ribosomal protein S28 MRPS28 HSPC007,
MRP-S28,
MRP-S35,
MRPS35
-34.2kb Sense Chr. 8 - 79918860 - 80030271 on NCBI Build GRCh38
Downstream microRNA 5708 MIR5708 70.2kb Anti-sense Chr. 8 - 80241389 - 80241473 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

anatomical structure morphogenesis
B cell differentiation
secretion
calcium ion binding
protein binding
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0