This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402255_m1
See other LOC100128006 GE Assays ›
Gene Symbol:
LOC100128006
Gene Name:
uncharacterized LOC100128006
Gene Aliases:
-
Chromosome Location:
Chr.17: 12760134 - 12790284 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402255_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100128006
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104607.1 1 - 2 135 71
GenBank mRNA BX649083.1 - 1 - 2 94 71
DA231808.1 - 1 - 2 136 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928418 LOC101928418 -54.0kb Sense Chr. 17 - 12671862 - 12706135 on NCBI Build GRCh38
Downstream microRNA 1269b MIR1269B 127.0kb Sense Chr. 17 - 12917268 - 12917342 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700009790.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176521.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176521.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176521.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176521.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176521.hg.1 TC1700009790.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700176523.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176523.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176523.hg.1 TC1700009790.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700176520.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176520.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176520.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176520.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176520.hg.1 TC1700009790.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009790.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay TC1700009790.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176522.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176522.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176522.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176522.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176522.hg.1 TC1700009790.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700176519.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176519.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176519.hg.1 ENST00000584772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700176519.hg.1 NR_104607.1 Refseq
Affymetrix Human Clariom D Assay PSR1700176519.hg.1 TC1700009790.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0