This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400414_m1
See other RSPO2 GE Assays ›
Gene Symbol:
RSPO2
Gene Name:
R-spondin 2
Gene Aliases:
CRISTIN2
Chromosome Location:
Chr.8: 107899316 - 108083685 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400414_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RSPO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282863.1 NP_001269792.1 2 - 3 542 105
GenBank mRNA BC036554.1 - 2 - 3 543 5265957 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011452.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
Wnt signaling pathway
positive regulation of Wnt signaling pathway
bone mineralization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of odontogenesis of dentin-containing tooth
lung growth
epithelial tube branching involved in lung morphogenesis
trachea cartilage morphogenesis
dopaminergic neuron differentiation
positive regulation of canonical Wnt signaling pathway
receptor binding
protein binding
heparin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay PSR0800177927.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177927.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay PSR0800177927.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177927.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay PSR0800177927.hg.1 TC0800011452.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay PSR0800177916.hg.1 TC0800011452.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000276659.9 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517781.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000517939.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000520026.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521502.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521757.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000521956.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 ENST00000522333.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_001282863.1 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq
Affymetrix Human Clariom D Assay TC0800011452.hg.1 NM_178565.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0