Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000397510.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000397512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000488769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000492023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000526170.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000608174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001042463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001276253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001276274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_174940.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000397510.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000397512.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000488769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000492023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000526170.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
ENST00000608174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001042463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001276253.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_001276274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
NM_174940.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144794.hg.1 |
TC1100006485.hg.1 |
Clariom D TC |