This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04398436_m1
See other TRIB3 GE Assays ›
Gene Symbol:
TRIB3
Gene Name:
tribbles pseudokinase 3
Gene Aliases:
C20orf97, NIPK, SINK, SKIP3, TRB3
Chromosome Location:
Chr.20: 380629 - 397559 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04398436_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRIB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301201.1 NP_001288130.1 1 - 2 209 72
GenBank mRNA AK297546.1 - 1 - 2 207 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK297546.1, NM_001301201.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
regulation of glucose transport
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
response to endoplasmic reticulum stress
regulation of MAP kinase activity
negative regulation of fat cell differentiation
negative regulation of fatty acid biosynthetic process
negative regulation of transcription, DNA-templated
positive regulation of ubiquitin-protein transferase activity
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
transcription corepressor activity
protein kinase inhibitor activity
protein binding
ATP binding
kinase activity
protein kinase binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000217233.7 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000422053.3 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000449710.5 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 ENST00000615226.4 Ensembl
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301188.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301190.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301193.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301196.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_001301201.1 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq
Affymetrix Human Clariom D Assay TC2000006444.hg.1 NM_021158.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0