This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04333925_g1
See other IFITM5 GE Assays ›
Gene Symbol:
IFITM5
Gene Name:
interferon induced transmembrane protein 5
Gene Aliases:
BRIL, DSPA1, Hrmp1, OI5, fragilis4
Chromosome Location:
Chr.11: 298200 - 299526 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04333925_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFITM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025295.2 NP_001020466.1 1 - 2 221 93
GenBank mRNA BC150562.1 - 1 - 2 214 40127610 93
BC150563.1 - 1 - 2 214 40127611 93
CR747200.1 - 1 - 2 223 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013119.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003502.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFITM5

Gene Ontology Categories:

Function(s)

in utero embryonic development
response to biotic stimulus
bone mineralization
regulation of bone mineralization
bone morphogenesis

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11015372.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015372.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015372.hg.1 TC11003502.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015371.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015371.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015371.hg.1 TC11003502.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203123.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203123.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203123.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203123.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203123.hg.1 TC1100013119.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203124.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203124.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203124.hg.1 ENST00000382614.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100203124.hg.1 NM_001025295.2 Refseq
Affymetrix Human Clariom D Assay PSR1100203124.hg.1 TC1100013119.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 163015 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0