This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332665_m1
See other C18ORF61 GE Assays ›
Gene Symbol:
C18orf61
Gene Name:
uncharacterized LOC497259
Gene Aliases:
-
Chromosome Location:
Chr.18: 12200779 - 12224711 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332665_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C18orf61
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_049896.1 4 - 5 797 66
GenBank mRNA AI683092.1 - 1 - 2 66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI005149.1 - 1 - 2 Chr. 18 - 12218720 - 12223174 [-] on Build GRCh38 66
GenBankmRNA BX104712.1 - 1 - 2 Chr. 18 - 12218769 - 12223178 [-] on Build GRCh38 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ankyrin repeat domain 62 ANKRD62 DKFZp779B1634 -88.9kb Sense Chr. 18 - 12093796 - 12181684 on NCBI Build GRCh38
Downstream cell death-inducing DFFA-like effector a CIDEA CIDE-A 29.6kb Sense Chr. 18 - 12254319 - 12277595 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800009217.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000583691.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000583691.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163292.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163292.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163292.hg.1 TC1800009217.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000579413.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000583691.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000583691.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008067.hg.1 ENST00000584734.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163289.hg.1 TC1800009217.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163290.hg.1 TC1800009217.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592203.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592203.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 ENST00000592801.5 Ensembl
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay TC1800009217.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000588436.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 ENST00000588744.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163288.hg.1 TC1800009217.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163291.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163291.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163291.hg.1 ENST00000586882.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163291.hg.1 NR_049896.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163291.hg.1 TC1800009217.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0