This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332495_m1
See other LINC00570 GE Assays ›
Gene Symbol:
LINC00570
Gene Name:
long intergenic non-protein coding RNA 570
Gene Aliases:
ncRNA-a5
Chromosome Location:
Chr.2: 11393981 - 11403077 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332495_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00570
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_047499.1 2 - 3 282 91
GenBank mRNA BX102688.1 - 2 - 3 282 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI493106.1 - 1 - 2 Chr. 2 - 11394110 - 11402246 [-] on Build GRCh38 91
GenBankmRNA N23720.1 - 1 - 2 Chr. 2 - 11399169 - 11402261 [-] on Build GRCh38 91
GenBankmRNA BU660565.1 - 3 - 4 417 Chr. 2 - 11398996 - 11402108 [+] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Rho associated coiled-coil containing protein kinase 2 ROCK2 ROCK-II -55.0kb Anti-sense Chr. 2 - 11179759 - 11345407 on NCBI Build GRCh38
Downstream E2F transcription factor 6 E2F6 E2F-6 41.3kb Anti-sense Chr. 2 - 11444375 - 11466177 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200006721.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146091.hg.1 TC0200006721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146099.hg.1 TC0200006721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146093.hg.1 TC0200006721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146092.hg.1 TC0200006721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay PSR0200146098.hg.1 TC0200006721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000417473.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000432665.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000437795.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 ENST00000453100.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq
Affymetrix Human Clariom D Assay TC0200006721.hg.1 NR_047499.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0