Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155426.hg.1 |
TC2100008189.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000440363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
ENST00000441910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008189.hg.1 |
NR_046774.2 |
Refseq |