This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332439_m1
See other DSCAM-IT1 GE Assays ›
Gene Symbol:
DSCAM-IT1
Gene Name:
DSCAM intronic transcript 1
Gene Aliases:
zag4
Chromosome Location:
Chr.21: 40618507 - 40630767 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332439_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSCAM-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046774.2 1 - 2 116 79
GenBank mRNA BG221591.1 - 106 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DSCAM antisense RNA 1 DSCAM-AS1 M41 -244.2kb Anti-sense Chr. 21 - 40383083 - 40385358 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 323 LINC00323 C21orf130,
NCRNA00323,
PRED42
510.7kb Sense Chr. 21 - 41141500 - 41148064 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008189.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay PSR2100155428.hg.1 TC2100008189.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay PSR2100155427.hg.1 TC2100008189.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000440363.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 ENST00000441910.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay TC2100008189.hg.1 NR_046774.2 Refseq
Affymetrix Human Clariom D Assay TC2100008189.hg.1 NR_046774.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0