This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332417_m1
See other PRICKLE2-AS2 GE Assays ›
Gene Symbol:
PRICKLE2-AS2
Gene Name:
PRICKLE2 antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 64103470 - 64106358 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332417_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRICKLE2-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046701.1 2 - 3 271 120
GenBank mRNA BX110661.1 - 2 - 3 271 120

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PRICKLE2 antisense RNA 1 PRICKLE2-AS1 -1.4kb Sense Chr. 3 - 64067964 - 64103131 on NCBI Build GRCh38
Downstream PRICKLE2 antisense RNA 3 PRICKLE2-AS3 81.6kb Sense Chr. 3 - 64187544 - 64201561 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007684.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155568.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155568.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155568.hg.1 TC0300007684.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007684.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay TC0300007684.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155565.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155565.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155565.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155565.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155565.hg.1 TC0300007684.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155566.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155566.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155566.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155566.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155566.hg.1 TC0300007684.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155567.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155567.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155567.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155567.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155567.hg.1 TC0300007684.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300155564.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155564.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155564.hg.1 ENST00000484703.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300155564.hg.1 NR_046701.1 Refseq
Affymetrix Human Clariom D Assay PSR0300155564.hg.1 TC0300007684.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0