This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332408_g1
See other ARHGAP26-AS1 GE Assays ›
Gene Symbol:
ARHGAP26-AS1
Gene Name:
ARHGAP26 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 142859604 - 142868910 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332408_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP26-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046680.1 5 - 6 841 72
GenBank mRNA BC039527.1 - 5 - 6 841 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG460080.1 - 1 - 2 Chr. 5 - 142859627 - 142866885 [+] on Build GRCh38 72
GenBankmRNA AI204525.1 - 1 - 2 Chr. 5 - 142859602 - 142860099 [+] on Build GRCh38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101926975 LOC101926975 -98.6kb Anti-sense Chr. 5 - 142745600 - 142760993 on NCBI Build GRCh38
Downstream ARHGAP26 intronic transcript 1 ARHGAP26-IT1 332.3kb Anti-sense Chr. 5 - 143192500 - 143194166 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012326.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185787.hg.1 TC0500012326.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000432015.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000433595.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 ENST00000434610.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq
Affymetrix Human Clariom D Assay TC0500012326.hg.1 NR_046680.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0