This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332394_m1
See other ZFX-AS1 GE Assays ›
Gene Symbol:
ZFX-AS1
Gene Name:
ZFX antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 24146225 - 24149654 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332394_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZFX-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046657.1 2 - 3 192 123
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW237142.1 - 1 - 2 Chr. X - 24146225 - 24149654 [+] on Build GRCh38 123

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 2 subunit gamma EIF2S3 EIF2,
EIF2G,
EIF2gamma,
eIF-2gA
-67.4kb Anti-sense Chr. X - 24054948 - 24078810 on NCBI Build GRCh38
Downstream zinc finger protein,
X-linked
ZFX ZNF926 92.0 Anti-sense Chr. X - 24148994 - 24216255 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009259.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009259.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00163612.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00163612.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00163612.hg.1 ENST00000427551.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00163612.hg.1 NR_046657.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00163612.hg.1 TC0X00009259.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0